Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsga10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290720.1 | NP_001277649.1 | 6, 7 | 941 | ||
NM_001290721.1 | NP_001277650.1 | 5, 6 | 748 | |||
NM_207228.3 | NP_997111.1 | 6, 7 | 808 | |||
GenBank mRNA | AF530050.3 | - | 5, 6 | 753 | ||
AK030254.1 | - | 6, 7 | 941 | |||
AK163138.1 | - | 6, 7 | 755 | |||
AK170053.1 | - | 6, 7 | 831 | |||
AY618264.1 | - | 5, 6 | 753 | |||
BC066782.1 | - | 6, 7 | 805 | 6705127 |