Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nomo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153057.4 | NP_694697.3 | 20, 21 | 2475 | ||
GenBank mRNA | AK030584.1 | - | 20, 21 | 2442 | ||
AK035957.1 | - | 20, 21 | 2447 | |||
AK080754.1 | - | 20, 21 | 2338 | |||
AK082495.1 | - | 20, 21 | 2354 | |||
AK082963.1 | - | 20, 21 | 2334 | |||
AK142338.1 | - | 20, 21 | 2445 | |||
AK150107.1 | - | 20, 21 | 2430 | |||
AK165643.1 | - | 20, 21 | 2338 | |||
AK166764.1 | - | 5, 6 | 485 | |||
AK203657.1 | - | 1, 2 | 92 | |||
AK213173.1 | - | 1, 2 | 92 | |||
BC022138.1 | - | 19, 20 | 2191 | |||
BC025133.1 | - | 3, 4 | 354 | |||
BC033923.1 | - | 9, 10 | 1059 | 5099899 | ||
BC072630.1 | - | 20, 21 | 2421 | 6833516 |