Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptchd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001093750.1 | NP_001087219.1 | 2 | 504 | ||
GenBank mRNA | AK036111.1 | - | 2 | 299 | ||
BC116312.1 | - | 2 | 502 | 40095445 | ||
BC116313.1 | - | 2 | 499 | 40095446 |