Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vstm2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290539.1 | NP_001277468.1 | 4 | 658 | ||
NM_145967.2 | NP_666079.2 | 4 | 658 | |||
GenBank mRNA | AK032473.1 | - | 4 | 657 | ||
AK079096.1 | - | 580 | ||||
AK137363.1 | - | 4 | 580 | |||
BC027127.1 | - | 4 | 610 | 5361611 | ||
BM963943.1 | - | 4 | 318 |