Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vstm2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145967.2 | NP_666079.2 | 5 | 980 | ||
GenBank mRNA | AK032473.1 | - | 5 | 979 | ||
AK134954.1 | - | 2 | 233 | |||
AK137363.1 | - | 5 | 902 | |||
BC027127.1 | - | 5 | 932 | 5361611 |