Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfyve26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008550.1 | NP_001008550.1 | 37 | 6941 | ||
GenBank mRNA | AK029679.1 | - | 5 | 704 | ||
AK042386.1 | - | 11 | 2284 | |||
AK169541.1 | - | 11 | 2282 | |||
AK220333.1 | - | 37 | 6912 | |||
BC141359.1 | - | 37 | 6924 | 9055971 |