Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178407.3 | NP_848494.2 | 10 | 2296 | ||
GenBank mRNA | AK037027.1 | - | 10 | 2205 |