Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapre2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162941.1 | NP_001156413.1 | 7 | 971 | ||
NM_001162942.1 | NP_001156414.1 | 7 | 914 | |||
NM_153058.4 | NP_694698.3 | 6 | 1022 | |||
GenBank mRNA | AK028823.1 | - | 7 | 972 | ||
AK038562.1 | - | 6 | 1009 | |||
AK040767.1 | - | 6 | 997 | |||
AK045493.1 | - | 6 | 905 | |||
AK045904.1 | - | 6 | 997 | |||
AK046095.1 | - | 6 | 965 | |||
AK046221.1 | - | 6 | 989 | |||
AK148954.1 | - | 6 | 997 | |||
BC025804.1 | - | 6 | 969 | 5101879 | ||
BC027056.1 | - | 6 | 988 | 5361238 | ||
BC028987.1 | - | 6 | 944 | 5362960 | ||
BC035254.1 | - | 6 | 974 | 5363176 |