Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam149a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153535.2 | NP_705763.1 | 7 | 1959 | ||
GenBank mRNA | AK034229.1 | - | 7 | 1186 | ||
AK038552.1 | - | 7 | 1174 | |||
AK140440.1 | - | 7 | 1959 | |||
AK141607.1 | - | 7 | 1902 | |||
AK220518.1 | - | 7 | 1195 | |||
BC035537.1 | - | 7 | 1850 | 5369152 |