Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rho | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145383.1 | NP_663358.1 | 3 | 725 | ||
GenBank mRNA | AK044333.1 | - | 3 | 745 | ||
AK044412.1 | - | 3 | 743 | |||
BC013125.1 | - | 3 | 725 | 4500760 | ||
BC031766.1 | - | 3 | 726 | 4527040 |
Set Membership: |
G-protein coupled receptor |