Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153391.2 | NP_700440.2 | 11 | 1082 | ||
GenBank mRNA | AK043836.1 | - | 11 | 1039 | ||
AK052303.1 | - | 11 | 1062 | |||
AK129413.1 | - | 3 | 171 | |||
AK147970.1 | - | 11 | 1082 | |||
AK166172.1 | - | 11 | 1048 | |||
AY029258.1 | - | 11 | 1152 | |||
BC060134.1 | - | 11 | 1076 | |||
BC082301.1 | - | 10 | 962 |