Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153391.2 | NP_700440.2 | 16, 17 | 1834 | ||
GenBank mRNA | AK052303.1 | - | 16, 17 | 1814 | ||
AK129413.1 | - | 8, 9 | 923 | |||
AK147970.1 | - | 16, 17 | 1834 | |||
AK166172.1 | - | 16, 17 | 1800 | |||
AY029258.1 | - | 16, 17 | 1904 | |||
BC046432.1 | - | 1, 2 | 36 | |||
BC060134.1 | - | 16, 17 | 1828 | |||
BC082301.1 | - | 15, 16 | 1714 |