Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdkl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166653.1 | NP_001160125.1 | 6 | 987 | ||
NM_001166654.1 | NP_001160126.1 | 6 | 1077 | |||
NM_001166655.1 | NP_001160127.1 | 6 | 1080 | |||
NM_001166656.1 | NP_001160128.1 | 6 | 987 | |||
NM_001166657.1 | NP_001160129.1 | 6 | 1077 | |||
NM_153785.4 | NP_722480.3 | 6 | 1077 | |||
GenBank mRNA | AK045036.1 | - | 5 | 1436 | ||
AK045356.1 | - | 6 | 977 | |||
AK046394.1 | - | 6 | 1078 | |||
AK050990.1 | - | 6 | 1077 | |||
AK078804.1 | - | 6 | 969 | |||
AK090045.1 | - | 6 | 863 | |||
AK166428.1 | - | 6 | 919 | |||
BC028871.1 | - | 6 | 949 | 4241862 |