Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pddc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172116.4 | NP_742114.3 | 7, 8 | 725 | ||
GenBank mRNA | AK081187.1 | - | 7, 8 | 685 | ||
AK084273.1 | - | 7, 8 | 681 | |||
AK088159.1 | - | 7, 8 | 700 | |||
AK140975.1 | - | 7, 8 | 676 | |||
AK150323.1 | - | 5, 6 | 1395 | |||
AK150354.1 | - | 5, 6 | 1395 | |||
AK151259.1 | - | 5, 6 | 1395 | |||
AK151794.1 | - | 5, 6 | 1395 | |||
AK151959.1 | - | 5, 6 | 1395 | |||
AK153110.1 | - | 5, 6 | 1395 | |||
AK161205.1 | - | 6, 7 | 1528 | |||
BC023835.1 | - | 7, 8 | 695 | 5345186 |