Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tacc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040435.2 | NP_001035525.1 | 12 | 2091 | ||
GenBank mRNA | AF093542.1 | - | 13 | 1325 | ||
AF156934.1 | - | 12 | 1645 | |||
AF203445.1 | - | 12 | 1592 | |||
AF203446.1 | - | 12 | 980 | |||
AF247674.1 | - | 13 | 1376 | |||
AK076282.1 | - | 2118 | ||||
AK145556.1 | - | 12 | 1652 | |||
AK160291.1 | - | 12 | 1654 | |||
BC003252.1 | - | 12 | 1648 | 3500413 | ||
BC055071.1 | - | 4067 | ||||
BC066184.1 | - | 12 | 1627 | 30457053 | ||
BC068314.1 | - | 12 | 1627 | 30461783 |