Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhdc8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144810.5 | NP_659059.1 | 1, 2 | 1015 | ||
GenBank mRNA | AK043039.1 | - | 1, 2 | 353 | ||
AK044318.1 | - | 1, 2 | 591 | |||
AK080356.1 | - | 1, 2 | 1015 | |||
BC011135.1 | - | 1, 2 | 580 | 4191503 | ||
BC023173.1 | - | 1, 2 | 214 |