Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr174 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033251.4 | NP_001028423.1 | 5 | 1412 | ||
NM_001177781.1 | NP_001171252.1 | 5 | 1573 | |||
NM_001177782.1 | NP_001171253.1 | 3 | 1317 | |||
GenBank mRNA | AK153611.1 | - | 5 | 1414 | ||
AK153950.1 | - | 4 | 1350 | |||
AK156069.1 | - | 5 | 1478 | |||
AK171832.1 | - | 5 | 1574 |
Set Membership: |
G-protein coupled receptor |