Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009315.3 | NP_033341.1 | 14 | 1684 | ||
GenBank mRNA | AK153731.1 | - | 1732 | |||
AK154867.1 | - | 14 | 1718 | |||
BC058583.1 | - | 14 | 1684 | 6514960 | ||
D49439.1 | - | 13 | 1617 |