Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam46d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163104.2 | NP_001156576.1 | 6 | 1253 | ||
NM_001271008.1 | NP_001257937.1 | 6 | 1149 | |||
GenBank mRNA | AK133246.1 | - | 6 | 1150 |