Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbbp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172517.2 | NP_766105.2 | 12 | 1331 | ||
GenBank mRNA | AK049247.1 | - | 12 | 1318 | ||
AK154782.1 | - | 12 | 1332 | |||
AK170520.1 | - | 12 | 1329 | |||
AK171177.1 | - | 12 | 1336 | |||
BC030892.1 | - | 6 | 532 | |||
BC057632.1 | - | 13 | 1204 | 4918318 |