Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172518.3 | NP_766106.2 | 9 | 1209 | ||
GenBank mRNA | AK028867.1 | - | 9 | 1055 | ||
AK150804.1 | - | 9 | 1200 | |||
AK151503.1 | - | 9 | 1166 | |||
AK165284.1 | - | 9 | 1183 | |||
AK170202.1 | - | 9 | 1211 | |||
AK171215.1 | - | 9 | 1212 | |||
AK173150.1 | - | 1 | 22 | |||
BC003960.1 | - | 6 | 450 | |||
BC058667.1 | - | 9 | 1200 | 6410012 |