Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tank | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164071.1 | NP_001157543.1 | 9 | 1582 | ||
NM_001164072.1 | NP_001157544.1 | 8 | 1374 | |||
NM_011529.2 | NP_035659.1 | 8 | 1373 | |||
GenBank mRNA | AK088817.1 | - | 8 | 1320 | ||
AK147727.1 | - | 9 | 1528 | |||
AK165420.1 | - | 8 | 1330 | |||
AK165994.1 | - | 10 | 1391 | |||
BC026148.1 | - | 8 | 1355 | 3492937 | ||
BC033282.1 | - | 10 | 1533 | |||
BC052685.1 | - | 8 | 2011 | |||
U51907.1 | - | 8 | 1312 | |||
U59864.1 | - | 8 | 1386 |