Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ythdf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145393.4 | NP_663368.3 | 4 | 716 | ||
GenBank mRNA | AK031616.1 | - | 4 | 587 | ||
AK034655.1 | - | 4 | 717 | |||
AK083882.1 | - | 4 | 515 | |||
AK144209.1 | - | 4 | 716 | |||
AK150350.1 | - | 4 | 570 | |||
AK150669.1 | - | 4 | 570 | |||
AK151392.1 | - | 4 | 570 | |||
AK151862.1 | - | 4 | 570 | |||
AK159548.1 | - | 4 | 573 | |||
BC014797.1 | - | 4 | 522 | 3962417 | ||
BC028994.1 | - | 4 | 517 | 5355331 |