Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cracr2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025103.2 | NP_001020274.2 | 2 | 227 | ||
GenBank mRNA | BC048916.1 | - | 2 | 227 | ||
BC098333.1 | - | 2 | 188 | 5256663 |