Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081230.1 | NP_001074699.1 | 9 | 1526 | ||
GenBank mRNA | AK030527.1 | - | 9 | 1367 | ||
AK135702.1 | - | 6 | 1020 | |||
AK135823.1 | - | 9 | 1430 | |||
AK138540.1 | - | 9 | 1471 | |||
AK147287.1 | - | 9 | 1469 | |||
AK147416.1 | - | 9 | 1469 | |||
AK162555.1 | - | 9 | 1446 | |||
BC051460.1 | - | 1 | 51 | |||
BC061216.1 | - | 9 | 1382 | |||
BC087935.1 | - | 9 | 1423 |