Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc44a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145394.3 | NP_663369.2 | 4 | 540 | ||
GenBank mRNA | AK145169.1 | - | 2 | 152 | ||
BC010552.1 | - | 3 | 308 | 3582543 | ||
BC025548.1 | - | 4 | 538 | 5320841 | ||
BY735109.1 | - | 2 | 152 | |||
CJ130557.1 | - | 1 | 41 |