Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172520.2 | NP_766108.1 | 3, 4 | 722 | ||
GenBank mRNA | AK031443.1 | - | 3, 4 | 723 | ||
AK053075.1 | - | 3, 4 | 723 | |||
BC060376.1 | - | 1, 2 | 452 | 5340007 |