Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013684.3 | NP_038712.3 | 7 | 1075 | ||
GenBank mRNA | AK085037.1 | - | 7 | 1076 | ||
BC016476.1 | - | 6 | 885 | 3584341 | ||
BC050136.1 | - | 7 | 1026 | 30020603 | ||
D01034.1 | - | 6 | 892 | |||
DQ906043.1 | - | 73 | ||||
U63933.1 | - | 8 | 1081 |