Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nutm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172521.1 | NP_766109.1 | 7 | 2942 | ||
GenBank mRNA | AK077049.1 | - | 7 | 2942 | ||
BC125332.1 | - | 7 | 2922 | 40129847 | ||
BC125334.1 | - | 7 | 2922 | 40129849 |