Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033253.3 | NP_001028425.1 | 6 | 866 | ||
NM_001159942.1 | NP_001153414.1 | 7 | 1032 | |||
GenBank mRNA | AK033676.1 | - | 7 | 1032 | ||
AK034403.1 | - | 7 | 1033 | |||
AK045140.1 | - | 7 | 1212 | |||
AK122468.1 | - | 3 | 190 | |||
AK135923.1 | - | 6 | 868 | |||
BC076628.1 | - | 4 | 263 | |||
BC117965.1 | - | 5 | 468 | 40098326 | ||
BC117966.1 | - | 5 | 461 | 40098330 |