Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbx15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009323.2 | NP_033349.2 | 8 | 1671 | ||
GenBank mRNA | AF041822.1 | - | 8 | 1571 | ||
AK132578.1 | - | 8 | 1672 | |||
AK137458.1 | - | 8 | 1465 | |||
AY662679.1 | - | 8 | 1604 |