Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bms1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194339.1 | NP_919320.1 | 21 | 3598 | ||
GenBank mRNA | AK129082.1 | - | 21 | 3558 | ||
AK133013.1 | - | 21 | 3548 | |||
AK162178.1 | - | 21 | 3658 | |||
BC030906.1 | - | 14 | 2398 | |||
BC057054.1 | - | 21 | 3598 | 6827225 |