Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col28a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037865.1 | NP_001032954.1 | 3 | 268 | ||
GenBank mRNA | AJ890449.1 | - | 3 | 268 | ||
BC145537.1 | - | 3 | 255 |