Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc18a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172523.3 | NP_766111.1 | 2, 3 | 271 | ||
GenBank mRNA | AJ555564.1 | - | 1, 2 | 135 | ||
AK033871.1 | - | 2, 3 | 227 | |||
AK034368.1 | - | 1, 2 | 492 | |||
AK035644.1 | - | 2, 3 | 254 | |||
AK041274.1 | - | 2, 3 | 223 | |||
BC078449.1 | - | 2, 3 | 186 | 5705966 |