Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc18a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172523.3 | NP_766111.1 | 3 | 541 | ||
GenBank mRNA | AJ555564.1 | - | 2 | 405 | ||
AK033871.1 | - | 3 | 497 | |||
AK034368.1 | - | 2 | 762 | |||
AK035644.1 | - | 3 | 524 | |||
AK041274.1 | - | 3 | 493 | |||
BC078449.1 | - | 3 | 456 | 5705966 |