Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc24a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144813.1 | NP_659062.1 | 6, 7 | 2397 | ||
GenBank mRNA | BC016094.1 | - | 6, 7 | 2397 | 4504496 | |
BC044859.1 | - | 5, 6 | 1833 |