Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfap2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025305.2 | NP_001020476.1 | 4, 5 | 1108 | ||
NM_001286340.1 | NP_001273269.1 | 4, 5 | 1073 | |||
NM_009334.3 | NP_033360.2 | 4, 5 | 1073 | |||
GenBank mRNA | AK005128.1 | - | 4, 5 | 1029 | ||
AK017373.1 | - | 4, 5 | 1075 | |||
AK028906.1 | - | 4, 5 | 1028 | |||
AK032045.1 | - | 4, 5 | 1110 | |||
X78197.1 | - | 4, 5 | 999 |