Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164147.1 | NP_001157619.1 | 11 | 1223 | ||
NM_001164148.1 | NP_001157620.1 | 11 | 1223 | |||
NM_001164149.1 | NP_001157621.1 | 11 | 1220 | |||
NM_001164150.1 | NP_001157622.1 | 11 | 1223 | |||
NM_001164151.1 | NP_001157623.1 | 11 | 1220 | |||
NM_001164152.1 | NP_001157624.1 | 11 | 1223 | |||
NM_001164153.1 | NP_001157625.1 | 11 | 1220 | |||
NM_011548.4 | NP_035678.3 | 11 | 1220 | |||
GenBank mRNA | AF352579.1 | - | 11 | 915 | ||
AK017617.1 | - | 11 | 1188 | |||
AK034777.1 | - | 11 | 1203 | |||
AK038738.1 | - | 11 | 1207 | |||
AK146786.1 | - | 11 | 1214 | |||
AK147735.1 | - | 10 | 879 | |||
AK156265.1 | - | 11 | 1203 | |||
AK161009.1 | - | 11 | 1209 | |||
AK170674.1 | - | 11 | 1218 | |||
BC006860.1 | - | 11 | 1174 | 2631291 | ||
BC018260.1 | - | 10 | 848 | 4187386 |