Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146048.1 | NP_001139520.1 | 4 | 656 | ||
NM_172528.3 | NP_766116.3 | 4 | 304 | |||
GenBank mRNA | AK051043.1 | - | 4 | 655 | ||
AK082721.1 | - | 4 | 305 | |||
AK147075.1 | - | 4 | 656 | |||
AK220166.1 | - | 4 | 674 | |||
BC046591.1 | - | 4 | 643 | |||
BC087542.1 | - | 4 | 503 | 4934980 |