Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk5rap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145990.3 | NP_666102.2 | 20 | 2331 | ||
GenBank mRNA | AK129411.1 | - | 20 | 2331 | ||
BC059253.1 | - | 8 | 922 | |||
BC079888.1 | - | 17 | 1975 | |||
BC086696.1 | - | 20 | 2233 | 30691910 |