Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fnbp1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114665.2 | NP_001108137.1 | 7, 8 | 789 | ||
NM_153118.3 | NP_694758.2 | 7, 8 | 789 | |||
GenBank mRNA | BC034510.1 | - | 2, 3 | 237 | 4481144 |