Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmprss4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145403.2 | NP_663378.1 | 9 | 1068 | ||
GenBank mRNA | AK078890.1 | - | 9 | 979 | ||
AY043240.1 | - | 9 | 1068 | |||
BC021368.1 | - | 9 | 1017 | 5030266 |