Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: L3mbtl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289711.1 | NP_001276640.1 | 5 | 695 | ||
NM_001289712.1 | NP_001276641.1 | 5 | 695 | |||
NM_145993.5 | NP_666105.2 | 5 | 695 | |||
GenBank mRNA | AK029115.1 | - | 5 | 640 | ||
AK036510.1 | - | 5 | 648 | |||
AK132012.1 | - | 5 | 648 | |||
AK141810.1 | - | 4 | 1829 | |||
AK145560.1 | - | 5 | 637 | |||
AK146632.1 | - | 5 | 651 | |||
AK156236.1 | - | 5 | 650 | |||
AY237000.1 | - | 5 | 628 | |||
AY237001.1 | - | 5 | 628 | |||
AY237002.1 | - | 5 | 628 | |||
BC023933.1 | - | 5 | 636 | 5325131 | ||
BC030864.1 | - | 5 | 598 | 4211489 |