Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iqub | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172535.3 | NP_766123.2 | 2 | 456 | ||
GenBank mRNA | AK029931.1 | - | 2 | 446 | ||
AK133019.1 | - | 2 | 437 | |||
BC137937.1 | - | 2 | 423 | 8860956 | ||
BC137957.1 | - | 2 | 423 | 8860976 |