Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp609 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172536.3 | NP_766124.2 | 2 | 432 | ||
GenBank mRNA | AK036715.1 | - | 1 | 165 | ||
BC016271.1 | - | 2 | 432 | |||
BC058943.1 | - | 1 | 337 | |||
BC150683.1 | - | 1 | 141 | 9087594 |