Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wfikkn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100454.1 | NP_001093924.1 | 2 | 2243 | ||
GenBank mRNA | BC026460.1 | - | 1 | 439 | ||
BC048715.1 | - | 3 | 1158 | |||
BC147298.1 | - | 2 | 1461 | 9056349 | ||
BC147299.1 | - | 2 | 1461 | 9056355 | ||
JQ080911.1 | - | 1 | 700 |