Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vezt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK031848.1 | - | 8 | 1236 | ||
AK036017.1 | - | 8 | 1173 | |||
AK042540.1 | - | 8 | 1167 | |||
BC056428.1 | - | 8 | 1160 | 5716245 |