Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178675.4 | NP_848790.2 | 2 | 718 | ||
GenBank mRNA | AK038601.1 | - | 2 | 713 | ||
AK048957.1 | - | 2 | 678 | |||
AK053426.1 | - | 2 | 687 | |||
AK053545.1 | - | 2 | 657 | |||
AK083768.1 | - | 2 | 657 | |||
AK139331.1 | - | 2 | 688 | |||
AK163806.1 | - | 2 | 518 | |||
AK165021.1 | - | 2 | 672 | |||
AK166268.1 | - | 2 | 645 | |||
AK207626.1 | - | 66 | ||||
BC059075.1 | - | 2 | 614 | 6826829 |