Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178675.4 | NP_848790.2 | 7 | 1369 | ||
GenBank mRNA | AK038601.1 | - | 7 | 1364 | ||
AK048957.1 | - | 7 | 1329 | |||
AK053426.1 | - | 7 | 1338 | |||
AK053545.1 | - | 7 | 1308 | |||
AK083768.1 | - | 7 | 1308 | |||
AK139331.1 | - | 7 | 1339 | |||
AK163806.1 | - | 7 | 1169 | |||
AK165021.1 | - | 7 | 1323 | |||
AK166268.1 | - | 7 | 1297 | |||
BC059075.1 | - | 7 | 1265 | 6826829 |