Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Traf3ip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153137.4 | NP_694777.3 | 8 | 1040 | ||
GenBank mRNA | AK031400.1 | - | 8 | 980 | ||
AK036608.1 | - | 6 | 1170 | |||
AK161211.1 | - | 5 | 650 | |||
AY383616.1 | - | 6 | 624 | |||
BC036561.1 | - | 8 | 848 | |||
BC137605.1 | - | 6 | 651 | 8860624 | ||
BC171933.1 | - | 6 | 651 | 9054217 |